missing translation for 'onlineSavingsMsg'
Learn More

4-Bromocatechol 98.0+%, TCI America™

Catalog No. p-7119525
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21731G 1 g
B21735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21731G Supplier TCI America Supplier No. B21731G

Chemical Identifiers

CAS 17345-77-6
Molecular Formula C6H5BrO2
Molecular Weight (g/mol) 189.008
MDL Number MFCD00869769
InChI Key AQVKHRQMAUJBBP-UHFFFAOYSA-N
Synonym 4-Bromo-1,2-dihydroxybenzene
PubChem CID 28487
ChEBI CHEBI:34388
IUPAC Name 4-bromobenzene-1,2-diol
SMILES C1=CC(=C(C=C1Br)O)O

Specifications

Melting Point 87°C
Quantity 1 g
Formula Weight 189.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromocatechol