missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzyl alcohol, 99%

Catalog No. AAA1319718
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1319718 50 g
AAA1319709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1319718 Supplier Thermo Scientific Chemicals Supplier No. A1319718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 873-75-6 | C7H7BrO | 187.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 873-75-6
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00004650
InChI Key VEDDBHYQWFOITD-UHFFFAOYSA-N
Synonym 4-bromobenzyl alcohol, 4-bromophenyl methanol, p-bromobenzyl alcohol, benzenemethanol, 4-bromo, para-bromobenzyl alcohol, 4-bromobenzylalcohol, p-bromo-benzyl alcohol, ccris 5119, 4-bromophenyl-methanol, benzyl alcohol, p-bromo
PubChem CID 70119
IUPAC Name (4-bromophenyl)methanol
SMILES C1=CC(=CC=C1CO)Br

Specifications

Melting Point 76°C to 79°C
Quantity 50 g
Beilstein 1931620
Formula Weight 187.04
Percent Purity 99%
Chemical Name or Material 4-Bromobenzyl alcohol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-851-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only