missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzenediazonium Tetrafluoroborate 97.0+%, TCI America™

Catalog No. B31145G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B31145G Supplier TCI America Supplier No. B31145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 673-40-5
Molecular Formula C6H4BBrF4N2
Molecular Weight (g/mol) 270.82
MDL Number MFCD00011894
InChI Key FXTCQUCYDJZGGU-UHFFFAOYSA-N
Synonym 4-bromobenzenediazonium tetrafluoroborate, acmc-209nyn, 4-bromobenzene-1-diazonium; tetrafluoroboranuide, boron +3 cation; 4-bromobenzenediazonium; tetrafluoride
PubChem CID 2734810
IUPAC Name 4-bromobenzene-1-diazonium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.BrC1=CC=C(C=C1)[N+]#N

Specifications

Melting Point 133°C
Quantity 5 g
UN Number 1759
Formula Weight 270.82
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromobenzenediazonium Tetrafluoroborate
RTECSNumber BQ9625000
TSCA No