missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoanisole, 98%, Thermo Scientific™

Catalog No. p-71686
Change view
Click to view available options
Quantity:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC106631000 100mL
AC106635000 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106631000 Supplier Thermo Scientific Chemicals Supplier No. 106631000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 104-92-7
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00000097
InChI Key QJPJQTDYNZXKQF-UHFFFAOYSA-N
Synonym 4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide
PubChem CID 7730
ChEBI CHEBI:47257
IUPAC Name 1-bromo-4-methoxybenzene
SMILES COC1=CC=C(C=C1)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.4900g/mL
Boiling Point 223.0°C
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5630 to 1.5650
Linear Formula BrC6H4OCH3
Quantity 100mL
Beilstein 06, 199
Specific Gravity 1.49
Merck Index 10, 1400
Solubility Information Solubility in water: immiscible
Formula Weight 187.04
Percent Purity 98%
Chemical Name or Material 4-Bromoanisole, 98%
Show More Show Less
EINECSNumber 203-252-1
RTECSNumber BZ8501000
TSCA TSCA

RUO – Research Use Only