missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N-methylaniline 95.0+%, TCI America™

Catalog No. B44035G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B44035G Supplier TCI America Supplier No. B44035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6911-87-1
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD03030391
InChI Key AYVPVDWQZAAZCM-UHFFFAOYSA-N
Synonym 4-bromo-n-methylbenzenamine, benzenamine, 4-bromo-n-methyl, n-methyl-4-bromoaniline, benzenamine,4-bromo-n-methyl, 4bromo-n-methylaniline, 4-bromo-n-methyl-aniline, n-methyl-4-bromo-aniline, acmc-1cfc7, 4-bromanyl-n-methyl-aniline, 4-bromo-phenyl-methyl-amine
PubChem CID 2757052
IUPAC Name 4-bromo-N-methylaniline
SMILES CNC1=CC=C(Br)C=C1

Specifications

Melting Point 11°C
Boiling Point 260°C
Quantity 5 g
Formula Weight 186.05
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-N-methylaniline