missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-DL-mandelic Acid 98.0+%, TCI America™

Catalog No. B06095G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B06095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B06095G Supplier TCI America Supplier No. B06095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6940-50-7
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.05
MDL Number MFCD00004232
InChI Key BHZBRPQOYFDTAB-UHFFFAOYNA-N
Synonym 4-bromomandelic acid, 2-4-bromophenyl-2-hydroxyacetic acid, p-bromomandelic acid, p-bromo mandelic acid, 4-bromo-dl-mandelic acid, benzeneacetic acid, 4-bromo-.alpha.-hydroxy, 4-bromophenyl hydroxy acetic acid, 4-bromo-alpha-hydroxyphenylacetic acid, 4-bromo-hydroxybenzeneaceticacid, mandelic acid, p-bromo
PubChem CID 97930
IUPAC Name 2-(4-bromophenyl)-2-hydroxyacetic acid
SMILES OC(C(O)=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 5 g
Formula Weight 231.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-DL-mandelic Acid