missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-5-fluoro-2-methoxyaniline, 96%, Thermo Scientific Chemicals

Catalog No. AAH5939703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5939703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5939703 Supplier Thermo Scientific Chemicals Supplier No. H5939703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 330794-03-1 | Molecular Formula: C7H7BrFNO | Molecular Weight: 220.04 g/mol

4-Bromo-5-fluoro-2-methoxyaniline is used as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-5-fluoro-2-methoxyaniline is used as a laboratory reagent.

Notes
Store in a cool place. Keep the container tightly closed. Incompatible with strong-oxidizing agents, strong acids and strong bases.

Chemical Identifiers

CAS 330794-03-1
Molecular Formula C7H7BrFNO
Molecular Weight (g/mol) 220.04
MDL Number MFCD21603958
InChI Key WOFOKKKWMDHLEX-UHFFFAOYSA-N
Synonym benzenamine, 4-bromo-5-fluoro-2-methoxy
PubChem CID 22347298
IUPAC Name 4-bromo-5-fluoro-2-methoxyaniline
SMILES COC1=CC(Br)=C(F)C=C1N

Specifications

Quantity 1 g
UN Number 2811
Formula Weight 220.04
Percent Purity 96%
Chemical Name or Material 4-Bromo-5-fluoro-2-methoxyaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only