missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-4',4″-dimethoxytriphenylamine 98.0+%, TCI America™

Catalog No. B46671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46671G 1 g
B46675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46671G Supplier TCI America Supplier No. B46671G

Chemical Identifiers

CAS 194416-45-0
Molecular Formula C20H18BrNO2
Molecular Weight (g/mol) 384.273
MDL Number MFCD22124385
InChI Key XXCDCFFPMMCPEE-UHFFFAOYSA-N
Synonym 4-Bromo-N,N-bis(4-methoxyphenyl)aniline
PubChem CID 10596161
IUPAC Name N-(4-bromophenyl)-4-methoxy-N-(4-methoxyphenyl)aniline
SMILES COC1=CC=C(C=C1)N(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)Br

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 1 g
Formula Weight 384.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-4′,4′′-dimethoxytriphenylamine