missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3-methylphenol 98.0+%, TCI America™
Supplier: TCI America B365825G
Specifications
| 4-Bromo-3-methylphenol | |
| 68°C | |
| C7H7BrO | |
| 25 g | |
| GPOQODYGMUTOQL-UHFFFAOYSA-N | |
| 4-bromo-3-methylphenol | |
| 72857 | |
| ≥98.0% (GC) |
| 14472-14-1 | |
| 145°C | |
| MFCD00079723 | |
| 4-bromo-3-methyl-phenol, 4-bromo-m-cresol, phenol, 4-bromo-3-methyl, 4-bromo-3-methyl phenol, 4-bromo-5-methyl-phenol, 4-brom-3-methylphenol, zlchem 434, pubchem2201, 3-methyl-4-bromophenol, 2-bromo-5-hydroxytoluene | |
| CC1=CC(O)=CC=C1Br | |
| 187.04 | |
| 187.04 | |
| Crystalline Powder |
Chemical Identifiers
| 14472-14-1 | |
| 187.04 | |
| GPOQODYGMUTOQL-UHFFFAOYSA-N | |
| 72857 | |
| CC1=CC(O)=CC=C1Br |
| C7H7BrO | |
| MFCD00079723 | |
| 4-bromo-3-methyl-phenol, 4-bromo-m-cresol, phenol, 4-bromo-3-methyl, 4-bromo-3-methyl phenol, 4-bromo-5-methyl-phenol, 4-brom-3-methylphenol, zlchem 434, pubchem2201, 3-methyl-4-bromophenol, 2-bromo-5-hydroxytoluene | |
| 4-bromo-3-methylphenol |
Safety and Handling
TSCA : No