missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-méthylphénol 98,0+%, TCI America™

Numéro de catalogue. p-7123082
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B36585G 5 g
B365825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B36585G Fournisseur TCI America Code fournisseur B36585G

Identifiants chimiques

CAS 14472-14-1
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00079723
InChI Key GPOQODYGMUTOQL-UHFFFAOYSA-N
Synonym 4-bromo-3-methyl-phenol, 4-bromo-m-cresol, phenol, 4-bromo-3-methyl, 4-bromo-3-methyl phenol, 4-bromo-5-methyl-phenol, 4-brom-3-methylphenol, zlchem 434, pubchem2201, 3-methyl-4-bromophenol, 2-bromo-5-hydroxytoluene
PubChem CID 72857
IUPAC Name 4-bromo-3-méthylphénol
SMILES CC1=CC(O)=CC=C1Br

Spécifications

Melting Point 68°C
Boiling Point 145°C
Quantity 5 g
Formula Weight 187.04
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material 4-Bromo-3-methylphenol