missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3-methylbenzoic Acid 97.0+%, TCI America™
Supplier: TCI America B31135G
Specifications
| 4-Bromo-3-methylbenzoic Acid | |
| 215°C | |
| C8H6BrO2 | |
| 5 g | |
| KWVXDZLVCISXIB-UHFFFAOYSA-M | |
| 4-bromo-3-methylbenzoate | |
| 82131 | |
| ≥97.0% (GC,T) |
| 7697-28-1 | |
| White-Yellow | |
| MFCD00039526 | |
| 4-bromo-m-toluic acid, 4-bromo-3-methyl-benzoic acid, 4-bromo-3-methylbenzoicacid, 3-methyl-4-bromobenzoic acid, 4-bromo-3-methyl benzoic acid, benzoic acid, 4-bromo-3-methyl, pubchem3807, 4bromo-3-methylbenzoic acid, 4-bromo-5-methylbenzoic acid, ksc379g0b | |
| CC1=CC(=CC=C1Br)C([O-])=O | |
| 214.04 | |
| 215.05 | |
| Crystalline Powder |
Chemical Identifiers
| 7697-28-1 | |
| 214.04 | |
| KWVXDZLVCISXIB-UHFFFAOYSA-M | |
| 82131 | |
| CC1=CC(=CC=C1Br)C([O-])=O |
| C8H6BrO2 | |
| MFCD00039526 | |
| 4-bromo-m-toluic acid, 4-bromo-3-methyl-benzoic acid, 4-bromo-3-methylbenzoicacid, 3-methyl-4-bromobenzoic acid, 4-bromo-3-methyl benzoic acid, benzoic acid, 4-bromo-3-methyl, pubchem3807, 4bromo-3-methylbenzoic acid, 4-bromo-5-methylbenzoic acid, ksc379g0b | |
| 4-bromo-3-methylbenzoate |
Safety and Handling
TSCA : No