missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylbenzoic Acid 97.0+%, TCI America™

Catalog No. p-7119961
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31135G 5 g
B311325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31135G Supplier TCI America Supplier No. B31135G
Only null left

Chemical Identifiers

CAS 7697-28-1
Molecular Formula C8H6BrO2
Molecular Weight (g/mol) 214.04
MDL Number MFCD00039526
InChI Key KWVXDZLVCISXIB-UHFFFAOYSA-M
Synonym 4-bromo-m-toluic acid, 4-bromo-3-methyl-benzoic acid, 4-bromo-3-methylbenzoicacid, 3-methyl-4-bromobenzoic acid, 4-bromo-3-methyl benzoic acid, benzoic acid, 4-bromo-3-methyl, pubchem3807, 4bromo-3-methylbenzoic acid, 4-bromo-5-methylbenzoic acid, ksc379g0b
PubChem CID 82131
IUPAC Name 4-bromo-3-methylbenzoate
SMILES CC1=CC(=CC=C1Br)C([O-])=O

Specifications

Melting Point 215°C
Color White-Yellow
Quantity 5 g
Formula Weight 215.05
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-methylbenzoic Acid