accessibility menu, dialog, popup

UserName

CH3BrOO

CAS RN 7697-28-1

IUPAC Name: 4-bromo-3-methylbenzoate
Synonyms: 4-bromo-3-methylbenzoate
Molecular Weight (g/mol): 214.04
Molecular Formula: C8H6BrO2
InChi Key: KWVXDZLVCISXIB-UHFFFAOYSA-M
SMILES: CC1=CC(=CC=C1Br)C([O-])=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)