missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-chloro-2-fluoropyridine, 98%

Catalog No. AAH64362MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64362MD 250 mg
AAH6436203 1 g
AAH6436206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64362MD Supplier Thermo Scientific Chemicals Supplier No. H64362MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1017793-21-3 | C5H2BrClFN | 210.43 g/mol

Useful intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 1017793-21-3
Molecular Formula C5H2BrClFN
Molecular Weight (g/mol) 210.43
MDL Number MFCD11040607
InChI Key UMQBGWKWOLRROL-UHFFFAOYSA-N
Synonym 4-bromo-3-chloro-2-fluoro-pyridine, pubchem22991, abbypharma ap-30-7506
PubChem CID 56604170
IUPAC Name 4-bromo-3-chloro-2-fluoropyridine
SMILES C1=CN=C(C(=C1Br)Cl)F

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 210.43
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Bromo-3-chloro-2-fluoropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only