missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dimethylpyrazole 98.0+%, TCI America™

Catalog No. p-7122863
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33065G 5 g
B330625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33065G Supplier TCI America Supplier No. B33065G

Chemical Identifiers

CAS 3398-16-1
Molecular Formula C5H7BrN2
Molecular Weight (g/mol) 175.029
MDL Number MFCD00005242
InChI Key RISOHYOEPYWKOB-UHFFFAOYSA-N
Synonym 4-bromo-3,5-dimethylpyrazole, pyrazole, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromopyrazole, 1h-pyrazole, 4-bromo-3,5-dimethyl, 3.5-dimethyl-4-bromopyrazole, acmc-1cnce, 5-23-05-00154 beilstein handbook reference, ksc495g9b, 4-bromo-3,5-dimethyl pyrazole, 4-bromo-3,5-dimethyl-pyrazole
PubChem CID 76937
IUPAC Name 4-bromo-3,5-dimethyl-1H-pyrazole
SMILES CC1=C(C(=NN1)C)Br

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 5 g
Formula Weight 175.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3,5-dimethylpyrazole
RTECSNumber UQ6210000
TSCA No