missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dihydroxybenzoic Acid 97.0+%, TCI America™

Catalog No. B142525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B142525G 25 g
B1425250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B142525G Supplier TCI America Supplier No. B142525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16534-12-6
Molecular Formula C7H5BrO4
Molecular Weight (g/mol) 233.017
MDL Number MFCD00002513
InChI Key NUTRHYYFCDEALP-UHFFFAOYSA-N
Synonym benzoic acid, 4-bromo-3,5-dihydroxy, 4-bromo-3,5-resorcylic acid, 4-bromo-alpha-resorcylic acid, 4-bromo-3,5-dihydroxybenzic acid, .alpha.-resorcylic acid, 4-bromo, 4-bromo-3,5-dihydroxy-benzoic acid, alpha-resorcylic acid, 4-bromo, pubchem8196, acmc-209dsh, ksc177s3r
PubChem CID 86023
IUPAC Name 4-bromo-3,5-dihydroxybenzoic acid
SMILES C1=C(C=C(C(=C1O)Br)O)C(=O)O

Specifications

Melting Point 273°C
Quantity 25 g
Formula Weight 233.02
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3,5-dihydroxybenzoic Acid
EINECSNumber (3)-3066&(3)-1572
TSCA Yes