missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dihydroxybenzoic acid, 97+%

Catalog No. AAA1891422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1891422 100 g
AAA1891414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1891422 Supplier Thermo Scientific Chemicals Supplier No. A1891422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16534-12-6 | C7H5BrO4 | 233.017 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16534-12-6
Molecular Formula C7H5BrO4
Molecular Weight (g/mol) 233.017
MDL Number MFCD00002513
InChI Key NUTRHYYFCDEALP-UHFFFAOYSA-N
Synonym benzoic acid, 4-bromo-3,5-dihydroxy, 4-bromo-3,5-resorcylic acid, 4-bromo-alpha-resorcylic acid, 4-bromo-3,5-dihydroxybenzic acid, .alpha.-resorcylic acid, 4-bromo, 4-bromo-3,5-dihydroxy-benzoic acid, alpha-resorcylic acid, 4-bromo, pubchem8196, acmc-209dsh, ksc177s3r
PubChem CID 86023
IUPAC Name 4-bromo-3,5-dihydroxybenzoic acid
SMILES C1=C(C=C(C(=C1O)Br)O)C(=O)O

Specifications

Melting Point 268°C to 272°C
Quantity 100 g
Beilstein 2368265
Sensitivity Light sensitive
Formula Weight 233.02
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-3,5-dihydroxybenzoic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 240-606-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only