missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-thiophenemethanol 98.0+%, TCI America™
Spécifications
Spécifications
| Color | White-Yellow |
| Quantity | 1 g |
| Synonym | 4-bromothiophen-2-yl methanol, 4-bromo-2-thienyl methanol, 4-bromo-2-hydroxymethylthiophene, 4-bromo-2-thiophenemethanol, 4-bromo-2-hydroxymethyl thiophene, 2-thiophenemethanol, 4-bromo, acmc-1bm1t, 3-bromo-5-thienyl methanol, 4-bromo-thiophene-2-methanol, 2-thiophenemethanol,4-bromo |
| Formula Weight | 193.06 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 4-Bromo-2-thiophenemethanol |
Sécurité et manipulation
| TSCA | No |