missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitrobenzaldehyde 96.0+%, TCI America™

Catalog No. B41461G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41461G 1 g
B41465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41461G Supplier TCI America Supplier No. B41461G

Chemical Identifiers

CAS 5551-12-2
Molecular Formula C7H4BrNO3
Molecular Weight (g/mol) 230.02
MDL Number MFCD00463687
InChI Key GSXUXSXBEUJRAJ-UHFFFAOYSA-N
PubChem CID 608099
IUPAC Name 4-bromo-2-nitrobenzaldehyde
SMILES C1=CC(=C(C=C1Br)[N+](=O)[O-])C=O

Specifications

Melting Point 96°C
Color Yellow
Quantity 1 g
Formula Weight 230.02
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-nitrobenzaldehyde