missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-hydroxypyridine 98.0+%, TCI America™

Catalog No. B43831G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B43831G Supplier TCI America Supplier No. B43831G

Chemical Identifiers

CAS 36953-37-4
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 173.997
MDL Number MFCD00234041
InChI Key SSLMGOKTIUIZLY-UHFFFAOYSA-N
Synonym 4-bromo-2-hydroxypyridine, 4-bromopyridin-2-ol, 4-bromopyridin-2 1h-one, 4-bromo-2-pyridinol, 4-bromo-1,2-dihydropyridin-2-one, 4-bromo-2 1h-pyridinone, 4-bromo-2-pyridone, 4-bromo-2-hydroxy pyridine, 2-hydroxy-4-bromopyridine, 2 1h-pyridinone, 4-bromo
PubChem CID 817115
IUPAC Name 4-bromo-1H-pyridin-2-one
SMILES C1=CNC(=O)C=C1Br

Specifications

Melting Point 209°C
Quantity 1 g
Formula Weight 174.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-hydroxypyridine