missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Bromo-2'-hydroxyacétophénone 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 42°C |
| Boiling Point | 83°C |
| Quantity | 1 g |
| Formula Weight | 215.05 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 4′-Bromo-2′-hydroxyacetophenone |
Sécurité et manipulation
| TSCA | No |