missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzamide, 96%

Catalog No. AAH52336MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52336MD 250 mg
AAH5233603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52336MD Supplier Thermo Scientific Chemicals Supplier No. H52336MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 292621-45-5 | C7H5BrFNO | 218.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 292621-45-5
Molecular Formula C7H5BrFNO
Molecular Weight (g/mol) 218.025
MDL Number MFCD03094432
InChI Key MJDRFCPNHLHNON-UHFFFAOYSA-N
Synonym acmc-209h7t, 4-bromo-2-fluoro-benzamide, benzamide,4-bromo-2-fluoro, benzamide, 4-bromo-2-fluoro
PubChem CID 2773331
IUPAC Name 4-bromo-2-fluorobenzamide
SMILES C1=CC(=C(C=C1Br)F)C(=O)N

Specifications

Melting Point 151°C to 153°C
Quantity 250 mg
Formula Weight 218.03
Percent Purity 96%
Chemical Name or Material 4-Bromo-2-fluorobenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only