missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluoroaniline 98.0+%, TCI America™

Catalog No. B131425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B131425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B131425G Supplier TCI America Supplier No. B131425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 367-24-8
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD00010221
InChI Key GZRMNMGWNKSANY-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromoaniline, benzenamine, 4-bromo-2-fluoro, 4-bromo-2-fluoro-phenylamine, 4-bromo-2-fluoro-aniline, 4-bromo-2-fluorobenzenamine, 4-bromo-2-fluoro aniline, 4-bromo-2-fluorophenylamine, 4-brom-2-fluoranilin, zlchem 545, pubchem1525
PubChem CID 123050
IUPAC Name 4-bromo-2-fluoroaniline
SMILES C1=CC(=C(C=C1Br)F)N

Specifications

Melting Point 40°C
Color White-Yellow
Boiling Point 122°C
Quantity 25 g
Formula Weight 190.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-fluoroaniline