missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chlorophenol 97.0+%, TCI America™

Catalog No. B18195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18195G 5 g
B181925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18195G Supplier TCI America Supplier No. B18195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3964-56-5
Molecular Formula C6H4BrClO
Molecular Weight (g/mol) 207.45
MDL Number MFCD00002166
InChI Key VIBJPUXLAKVICD-UHFFFAOYSA-N
Synonym 2-chloro-4-bromophenol, phenol, 4-bromo-2-chloro, phenol, 4-bromochloro, unii-994sj34d8p, 4-bromo-2-chloro-phenol, attercop-chm at106179, 4-bromo-2-chlorophenol 25gr, 4-bromo-chlorophenol, pubchem3596, acmc-209j6p
PubChem CID 19859
ChEBI CHEBI:38852
IUPAC Name 4-bromo-2-chlorophenol
SMILES OC1=CC=C(Br)C=C1Cl

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 234°C
Quantity 5 g
Formula Weight 207.45
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-chlorophenol