missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chlorobenzaldehyde 97.0+%, TCI America™

Numéro de catalogue. B39485G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B39485G 5 g
B39481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B39485G Fournisseur TCI America Code fournisseur B39485G
Il en reste null

Identifiants chimiques

CAS 158435-41-7
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.46
MDL Number MFCD08741413
InChI Key DHGPLNJITGVCSG-UHFFFAOYSA-N
PubChem CID 14109109
IUPAC Name 4-bromo-2-chlorobenzaldehyde
SMILES ClC1=C(C=O)C=CC(Br)=C1

Spécifications

Melting Point 75°C
Color White-Yellow
Quantity 5 g
Synonym 2-chloro-4-bromobenzaldehyde, 4-bromo-2-chloro-benzaldehyde, benzaldehyde, 4-bromo-2-chloro, acmc-1c7dt, intermediates-zcf02199, ksc497g8p, benzaldehyde,4-bromo-2-chloro, 4-bromanyl-2-chloranyl-benzaldehyde, 2-chloro-4-bromobenzaldehyde, 95+%
Formula Weight 219.46
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-chlorobenzaldehyde