missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-2'-chloroacetanilide 98.0+%, TCI America™

Catalog No. p-7119661
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24135G 5 g
B241325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24135G Supplier TCI America Supplier No. B24135G

Chemical Identifiers

CAS 3460-23-9
Molecular Formula C8H7BrClNO
Molecular Weight (g/mol) 248.504
MDL Number MFCD00040852
InChI Key MITWNEIUIPGZKR-UHFFFAOYSA-N
Synonym 4'-bromo-2'-chloroacetanilide, n-4-bromo-2-chlorophenyl acetamide, 4-bromo-2-chloroacetanilide, acetamide, n-4-bromo-2-chlorophenyl, maybridge3_007422, 2-chlor-4-bromacetanilid, acmc-209i8u, buttpark 83\07-60, acetanilide, 4'-bromo-2'-chloro
PubChem CID 610167
IUPAC Name N-(4-bromo-2-chlorophenyl)acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)Br)Cl

Specifications

Melting Point 151°C
Color White-Yellow
Quantity 5 g
Formula Weight 248.50
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromo-2′-chloroacetanilide