missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-chloro-1-iodobenzene 97.0+%, TCI America™
Supplier: TCI America B303025G
Specifications
| 4-Bromo-2-chloro-1-iodobenzene | |
| Yellow | |
| C6H3BrClI | |
| 25 g | |
| OHHKQBZOURGNLR-UHFFFAOYSA-N | |
| 4-bromo-2-chloro-1-iodobenzene | |
| 2735547 | |
| ≥97.0% (GC) |
| 31928-47-9 | |
| 104°C | |
| MFCD00079709 | |
| 4-bromo-2-chloroiodobenzene, 1-bromo-3-chloro-4-iodobenzene, benzene, 4-bromo-2-chloro-1-iodo, buttpark 34\07-78, 4-bromo-2-chloro-1-iodo-benzene, pubchem3592, acmc-1cp6y, 3-chloro-4-iodobromobenzene, ksc493m0p, 4-bromo-2-chloro-iodobenzene | |
| C1=CC(=C(C=C1Br)Cl)I | |
| 317.348 | |
| 317.35 | |
| Crystalline Lumps |
Chemical Identifiers
| 31928-47-9 | |
| 317.348 | |
| OHHKQBZOURGNLR-UHFFFAOYSA-N | |
| 2735547 | |
| C1=CC(=C(C=C1Br)Cl)I |
| C6H3BrClI | |
| MFCD00079709 | |
| 4-bromo-2-chloroiodobenzene, 1-bromo-3-chloro-4-iodobenzene, benzene, 4-bromo-2-chloro-1-iodo, buttpark 34\07-78, 4-bromo-2-chloro-1-iodo-benzene, pubchem3592, acmc-1cp6y, 3-chloro-4-iodobromobenzene, ksc493m0p, 4-bromo-2-chloro-iodobenzene | |
| 4-bromo-2-chloro-1-iodobenzene |
Safety and Handling
TSCA : No