accessibility menu, dialog, popup

UserName

BrClI

CAS RN 31928-47-9

IUPAC Name: 4-bromo-2-chloro-1-iodobenzene
Synonyms: 4-bromo-2-chloro-1-iodobenzene
Molecular Weight (g/mol): 317.35
Molecular Formula: C6H3BrClI
InChi Key: OHHKQBZOURGNLR-UHFFFAOYSA-N
SMILES: ClC1=C(I)C=CC(Br)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)