missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloro-1-iodobenzene 97.0+%, TCI America™

Catalog No. p-7119902
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30305G 5 g
B303025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30305G Supplier TCI America Supplier No. B30305G

Chemical Identifiers

CAS 31928-47-9
Molecular Formula C6H3BrClI
Molecular Weight (g/mol) 317.348
MDL Number MFCD00079709
InChI Key OHHKQBZOURGNLR-UHFFFAOYSA-N
Synonym 4-bromo-2-chloroiodobenzene, 1-bromo-3-chloro-4-iodobenzene, benzene, 4-bromo-2-chloro-1-iodo, buttpark 34\07-78, 4-bromo-2-chloro-1-iodo-benzene, pubchem3592, acmc-1cp6y, 3-chloro-4-iodobromobenzene, ksc493m0p, 4-bromo-2-chloro-iodobenzene
PubChem CID 2735547
IUPAC Name 4-bromo-2-chloro-1-iodobenzene
SMILES C1=CC(=C(C=C1Br)Cl)I

Specifications

Color Yellow
Boiling Point 104°C
Quantity 5 g
Formula Weight 317.35
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromo-2-chloro-1-iodobenzene