missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,5-dimethoxybenzaldehyde, 98+%

Catalog No. AAB2550006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2550006 5 g
AAB2550014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2550006 Supplier Thermo Scientific Chemicals Supplier No. B2550006

CAS: 31558-41-5 | C9H9BrO3 | 245.072 g/mol

4-Bromo-2,5-dimethoxybenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,5-dimethoxybenzene is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 31558-41-5
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.072
MDL Number MFCD00156964
InChI Key BIFWGDWGCZLCHF-UHFFFAOYSA-N
Synonym 2,5-dimethoxy-4-bromobenzaldehyde, pubchem2068, acmc-1aiq9, cbmicro_047429, cambridge id 6238031, 4-bromo-2,5-dimethoxy benzaldehyde, 4-bromo-2,5-dimethoxy-benzaldehyde, 4-bromanyl-2,5-dimethoxy-benzaldehyde, benzaldehyde, 4-bromo-2,5-dimethoxy
PubChem CID 777997
IUPAC Name 4-bromo-2,5-dimethoxybenzaldehyde
SMILES COC1=CC(=C(C=C1C=O)OC)Br

Specifications

Melting Point 132°C to 135°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 245.08
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-2,5-dimethoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only