missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,5-difluoroaniline 98.0+%, TCI America™

Catalog No. B20725G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20725G 5 g
B20721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20725G Supplier TCI America Supplier No. B20725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112279-60-4
Molecular Formula C6H4BrF2N
Molecular Weight (g/mol) 208.006
MDL Number MFCD01861125
InChI Key XOYHFIQPPOJMFK-UHFFFAOYSA-N
Synonym 4-bromo-2,5-difluoro-phenylamine, 4-bromo-2,5-difluoro-aniline, 4-bromo-2,5-difluorophenylamine, pubchem2928, acmc-1bvn4, ksc494m7l, 4-bromo-2,5-difluorobenzenamine, 2,5-difluoro-4-bromoaniline, 4-bromo-2,5-difluoroaniline, benzenamine, 4-bromo-2,5-difluoro
PubChem CID 2773285
IUPAC Name 4-bromo-2,5-difluoroaniline
SMILES C1=C(C(=CC(=C1F)Br)F)N

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 5 g
Formula Weight 208.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,5-difluoroaniline