missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1H-pyrazole, 98+%

Catalog No. AAH6467336
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6467336 500 g
AAH6467314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6467336 Supplier Thermo Scientific Chemicals Supplier No. H6467336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2075-45-8 | C3H3BrN2 | 146.98 g/mol

4-Bromopyrazole may be used in the preparation of 4-bromo-1-(2-chloroethyl)-1H-pyrazole. It may be used as starting material in the synthesis of 1,4?-bipyrazoles. Also used in the synthesis of various pharmaceutical and biologically active compounds, including inhibitors. It is also shown to be one of the 4-substituted pyrazoles, being able to act as inhibitor of liver alcohol dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromopyrazole may be used in the preparation of 4-bromo-1-(2-chloroethyl)-1H-pyrazole. It may be used as starting material in the synthesis of 1,4′-bipyrazoles. Also used in the synthesis of various pharmaceutical and biologically active compounds, including inhibitors. It is also shown to be one of the 4-substituted pyrazoles, being able to act as inhibitor of liver alcohol dehydrogenase.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 2075-45-8
Molecular Formula C3H3BrN2
Molecular Weight (g/mol) 146.98
MDL Number MFCD00075602
InChI Key WVGCPEDBFHEHEZ-UHFFFAOYSA-N
Synonym 4-bromopyrazole, 1h-pyrazole, 4-bromo, 4-brompyrazole, 4-bromo-pyrazole, pyrazole, 4-bromo, 4-bromo pyrazole, byz, zlchem 126, 1h-pyrazole,4-bromo, pubchem11041
PubChem CID 16375
IUPAC Name 4-bromo-1H-pyrazole
SMILES BrC1=CNN=C1

Specifications

Melting Point 93°C to 96°C (literature)
Boiling Point 250°C to 260°C (literature)
Quantity 500 g
Beilstein 106510
Solubility Information Slightly soluble in water.
Formula Weight 146.97
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material 4-Bromo-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UQ6200000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only