missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-indanone 97,0+%, TCI America™

Numéro de catalogue. B38011G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B38011G 1 g
B38015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B38011G Fournisseur TCI America Code fournisseur B38011G

Identifiants chimiques

CAS 15115-60-3
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.06
MDL Number MFCD01719772
InChI Key UVVYFYLSZIMKMC-UHFFFAOYSA-N
Synonym 4-bromo-1-indanone, 4-bromo-indan-1-one, 4-bromoindan-1-one, 4-bromo-2,3-dihydro-1h-inden-1-one, 1-indanone, 4-bromo, 4-bromohydrindone-1, 1h-inden-1-one, 4-bromo-2,3-dihydro, 4-bromoindanone, 4-bromo indanone, 4-bromo-indanone
PubChem CID 98713
IUPAC Name 4-bromo-2,3-dihydro-1H-inden-1-un
SMILES BrC1=CC=CC2=C1CCC2=O

Spécifications

Melting Point 94°C
Boiling Point 125°C
Quantity 1 g
Formula Weight 211.06
Percent Purity ≥97.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material 4-Bromo-1-indanone
RTECSNumber NK7536100
TSCA No