missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,2-phenylenediamine 96.0+%, TCI America™

Supplier:  TCI America B318125G

 View more versions of this product

Catalog No. B318125G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Bromo-1,2-phenylenediamine
64°C
MFCD02660622
4-bromo-1,2-benzenediamine, 4-bromo-1,2-diaminobenzene, 4-bromo-o-phenylenediamine, 1,2-diamino-4-bromobenzene, 2-amino-4-bromoaniline, 4-bromo-1,2-phenylenediamine, 4-bromo-2-aminoaniline, 1,2-benzenediamine, 4-bromo, 2-amino-4-bromophenylamine, 4-bromophenylene-1,2-diamine
NC1=CC=C(Br)C=C1N
187.04
187.04
Crystalline Powder
1575-37-7
C6H7BrN2
25 g
WIHHVKUARKTSBU-UHFFFAOYSA-N
4-bromobenzene-1,2-diamine
323593
≥96.0% (GC)

Chemical Identifiers

1575-37-7
187.04
WIHHVKUARKTSBU-UHFFFAOYSA-N
323593
NC1=CC=C(Br)C=C1N
C6H7BrN2
MFCD02660622
4-bromo-1,2-benzenediamine, 4-bromo-1,2-diaminobenzene, 4-bromo-o-phenylenediamine, 1,2-diamino-4-bromobenzene, 2-amino-4-bromoaniline, 4-bromo-1,2-phenylenediamine, 4-bromo-2-aminoaniline, 1,2-benzenediamine, 4-bromo, 2-amino-4-bromophenylamine, 4-bromophenylene-1,2-diamine
4-bromobenzene-1,2-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions