missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,2-phenylenediamine 96.0+%, TCI America™

Catalog No. p-7138109
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31815G 5 g
B318125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31815G Supplier TCI America Supplier No. B31815G

Chemical Identifiers

CAS 1575-37-7
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD02660622
InChI Key WIHHVKUARKTSBU-UHFFFAOYSA-N
Synonym 4-bromo-1,2-benzenediamine, 4-bromo-1,2-diaminobenzene, 4-bromo-o-phenylenediamine, 1,2-diamino-4-bromobenzene, 2-amino-4-bromoaniline, 4-bromo-1,2-phenylenediamine, 4-bromo-2-aminoaniline, 1,2-benzenediamine, 4-bromo, 2-amino-4-bromophenylamine, 4-bromophenylene-1,2-diamine
PubChem CID 323593
IUPAC Name 4-bromobenzene-1,2-diamine
SMILES NC1=CC=C(Br)C=C1N

Specifications

Melting Point 64°C
Quantity 5 g
Formula Weight 187.04
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-1,2-phenylenediamine