accessibility menu, dialog, popup

UserName

NH2BrH2N

CAS RN 1575-37-7

IUPAC Name: 4-bromobenzene-1,2-diamine
Synonyms: 4-bromobenzene-1,2-diamine
Molecular Weight (g/mol): 187.04
Molecular Formula: C6H7BrN2
InChi Key: WIHHVKUARKTSBU-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C=C1N
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)