missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenyl Benzoate 98.0+%, TCI America™

Catalog No. B18665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18665G 5 g
B186625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18665G Supplier TCI America Supplier No. B18665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2170-13-0
Molecular Formula C19H14O2
Molecular Weight (g/mol) 274.319
MDL Number MFCD00223541
InChI Key CINHWMYRCOGYIX-UHFFFAOYSA-N
Synonym Benzoic Acid 4-Biphenyl Ester
PubChem CID 231743
IUPAC Name (4-phenylphenyl) benzoate
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)OC(=O)C3=CC=CC=C3

Specifications

Melting Point 151°C
Color White
Quantity 5 g
Formula Weight 274.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Biphenyl Benzoate