missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. B177925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B177925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B177925G Supplier TCI America Supplier No. B177925G

Chemical Identifiers

CAS 6547-53-1
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.274
MDL Number MFCD00017540
InChI Key XJHGAJLIKDAOPE-UHFFFAOYSA-N
Synonym 4-benzyloxyphenylacetic acid, 2-4-benzyloxy phenyl acetic acid, 4-benzyloxy phenyl acetic acid, 4-benzyloxyphenyl acetic acid, benzeneacetic acid, 4-phenylmethoxy, 4-benzyloxy phenylacetic acid, 4-benzyloxy-phenyl-acetic acid, 2-4-phenylmethoxyphenyl acetic acid, 2-4-phenylmethoxy phenyl acetic acid
PubChem CID 81033
IUPAC Name 2-(4-phenylmethoxyphenyl)acetic acid
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(=O)O

Specifications

Melting Point 125°C
Color White
Quantity 25 g
Formula Weight 242.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxyphenylacetic Acid