missing translation for 'onlineSavingsMsg'
Learn More

(4-Benzyloxyphenyl)acetic acid, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 394080010

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC394080010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

6547-53-1
242.27
XJHGAJLIKDAOPE-UHFFFAOYSA-N
81033
C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(=O)O
C15H14O3
MFCD00017540
4-benzyloxyphenylacetic acid, 2-4-benzyloxy phenyl acetic acid, 4-benzyloxy phenyl acetic acid, 4-benzyloxyphenyl acetic acid, benzeneacetic acid, 4-phenylmethoxy, 4-benzyloxy phenylacetic acid, 4-benzyloxy-phenyl-acetic acid, 2-4-phenylmethoxyphenyl acetic acid, 2-4-phenylmethoxy phenyl acetic acid
2-(4-phenylmethoxyphenyl)acetic acid

Specifications

(4-Benzyloxyphenyl)acetic acid
98.5
Authentic
Glass bottle
C6H5CH2OC6H4CH2CO2H
MFCD00017540
XJHGAJLIKDAOPE-UHFFFAOYSA-N
2-(4-phenylmethoxyphenyl)acetic acid
81033
99%
6547-53-1
100.0
98.5% min. (HPLC)
C15H14O3
1g
4-benzyloxyphenylacetic acid, 2-4-benzyloxy phenyl acetic acid, 4-benzyloxy phenyl acetic acid, 4-benzyloxyphenyl acetic acid, benzeneacetic acid, 4-phenylmethoxy, 4-benzyloxy phenylacetic acid, 4-benzyloxy-phenyl-acetic acid, 2-4-phenylmethoxyphenyl acetic acid, 2-4-phenylmethoxy phenyl acetic acid
C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(=O)O
242.27
242.27
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 229-463-9

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...