missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyphenol, 98+%

Catalog No. AAA1484322
Change view
Click to view available options
Quantity:
25 g
100 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1484322 100 g
AAA1484314 25 g
AAA148430B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1484322 Supplier Thermo Scientific Chemicals Supplier No. A1484322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103-16-2 | C13H12O2 | 200.237 g/mol

4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.

Solubility
Slightly soluble in water. Soluble in alcohol, acetone, benzene, and diethyl ether.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 103-16-2
Molecular Formula C13H12O2
MDL Number MFCD00002333
InChI Key VYQNWZOUAUKGHI-UHFFFAOYSA-N
Synonym 4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon
PubChem CID 7638
ChEBI CHEBI:34380
IUPAC Name 4-phenylmethoxyphenol

Specifications

Melting Point 118°C to 122°C
Density 1.26
Quantity 100 g
Beilstein 1958305
Merck Index 14,6248
Solubility Information Slightly soluble in water. Soluble in alcohol,acetone,benzene,and diethyl ether.
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)O
Molecular Weight (g/mol) 200.237
Formula Weight 200.24
Percent Purity ≥98%
Chemical Name or Material 4-Benzyloxyphenol
Show More Show Less
Hazard Category H317-H319
Hazard Statement GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c
EINECSNumber 203-083-3
RTECSNumber SJ7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only