missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-2-hydroxybenzaldehyde, 99%

Catalog No. AAL1551506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1551506 5 g
AAL1551503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1551506 Supplier Thermo Scientific Chemicals Supplier No. L1551506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52085-14-0 | C14H12O3 | 228.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52085-14-0
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD01075698
InChI Key AMLKEDBYDOCGEG-UHFFFAOYSA-N
Synonym 4-benzyloxy-2-hydroxybenzaldehyde, benzaldehyde, 2-hydroxy-4-phenylmethoxy, 4-benzyloxy-2-hydroxy-benzaldehyde, 2-hydroxy-4-phenylmethoxy benzaldehyde, acmc-20amov, 4-benzyloxysalicylaldehyde, 4-benzyloxy-2-hydoxy-benzaldehyde, 4-bezyloxy-2-hydroxy-benzaldehyde, 4-benzyloxy-2-hydroxy benzaldehyde
PubChem CID 561235
IUPAC Name 2-hydroxy-4-phenylmethoxybenzaldehyde
SMILES C1=CC=C(C=C1)COC2=CC(=C(C=C2)C=O)O

Specifications

Melting Point 77°C to 80°C
Quantity 5 g
Beilstein 982995
Sensitivity Air Sensitive
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material 4-Benzyloxy-2-hydroxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only