missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-2-fluorobenzoic acid, 98%

Catalog No. AAH3148806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3148806 5 g
AAH3148803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3148806 Supplier Thermo Scientific Chemicals Supplier No. H3148806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 114045-96-4 | C14H11FO3 | 246.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114045-96-4
Molecular Formula C14H11FO3
Molecular Weight (g/mol) 246.24
MDL Number MFCD12407082
InChI Key HWQRXVRKZFDKLJ-UHFFFAOYSA-N
Synonym 4-benzyloxy-2-fluorobenzoic acid, benzoic acid, 2-fluoro-4-phenylmethoxy, acmc-20mjm1, 2-fluoro-4-benzyloxybenzoic acid, 4-benzyloxy-2-fluorobenzoicacid, 4-benzyloxy-2-fluoro-benzoic acid
PubChem CID 22314502
IUPAC Name 2-fluoro-4-phenylmethoxybenzoic acid
SMILES OC(=O)C1=C(F)C=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 165°C to 168°C
Quantity 5 g
Formula Weight 246.24
Percent Purity 98%
Chemical Name or Material 4-Benzyloxy-2-fluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only