missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl L-Aspartate 98.0+%, TCI America™

Catalog No. B32095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32095G 5 g
B320925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32095G Supplier TCI America Supplier No. B32095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2177-63-1
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.228
MDL Number MFCD00037208
InChI Key VGALFAWDSNRXJK-VIFPVBQESA-N
Synonym h-asp obzl-oh, l-aspartic acid 4-benzyl ester, l-aspartic acid beta-benzyl ester, beta-benzyl l-aspartate, 2s-2-amino-4-benzyloxy-4-oxobutanoic acid, 4-benzyl l-aspartate, benzyl hydrogen beta-l-aspartate, s-2-amino-4-benzyloxy-4-oxobutanoic acid, 2s-2-amino-4-oxo-4-phenylmethoxybutanoic acid, asp obzl
PubChem CID 101186
IUPAC Name (2S)-2-amino-4-oxo-4-phenylmethoxybutanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CC(C(=O)O)N

Specifications

Melting Point 221°C
Color White
Quantity 5 g
Formula Weight 223.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyl L-Aspartate