missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzoyl 4'-Methyldiphenyl Sulfide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 83°C |
| Color | Red-White |
| Quantity | 10 g |
| Formula Weight | 304.41 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Benzoyl 4′-Methyldiphenyl Sulfide |
Safety and Handling
| EINECSNumber | (4)-1779 |
| TSCA | Yes |