missing translation for 'onlineSavingsMsg'
Learn More

4-(Azidomethyl)benzeneboronic acid pinacol ester, 95%

Catalog No. p-7039228
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5840103 1 g
AAH5840106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5840103 Supplier Thermo Scientific Chemicals Supplier No. H5840103
Only null left

CAS: 1239481-05-0 | C13H18BN3O2 | 259.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with oxidizing agents. Store in a dry, cool conditions in a well sealed containers. Store at 4°C.

Chemical Identifiers

CAS 1239481-05-0
Molecular Formula C13H18BN3O2
Molecular Weight (g/mol) 259.116
MDL Number MFCD20922785
InChI Key CARNXRHOIGMOQD-UHFFFAOYSA-N
Synonym 2-4-azidomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, amtb503, 4-azidomethyl benzeneboronic acid pinacol ester
PubChem CID 46872199
IUPAC Name 2-[4-(azidomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN=[N+]=[N-]

Specifications

Quantity 1 g
Formula Weight 260.12
Percent Purity 95%
Chemical Name or Material 4-(Azidomethyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only