missing translation for 'onlineSavingsMsg'
Learn More

4-Aminostilbene 97.0+%, TCI America™

Catalog No. A08545G
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
A08545G 5 g
A085425G 25 g
2 options

Catalog No. A08545G

Supplier: TCI America A08545G

Only null left

Chemical Identifiers

CAS 834-24-2
Molecular Formula C14H13N
Molecular Weight (g/mol) 195.27
MDL Number MFCD00025377
InChI Key VFPLSXYJYAKZCT-VOTSOKGWSA-N
Synonym 4-Stilbenamine
PubChem CID 1272788
IUPAC Name 4-[(1E)-2-phenylethenyl]aniline
SMILES NC1=CC=C(\C=C\C2=CC=CC=C2)C=C1

Specifications

Melting Point 151°C
Color Yellow
Quantity 5 g
Formula Weight 195.27
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminostilbene
RTECSNumber WJ3500000
TSCA No