missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)benzonitrile Hydrochloride 98.0+%, TCI America™

Catalog No. A16455G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16455G 5 g
A16451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16455G Supplier TCI America Supplier No. A16455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15996-76-6
Molecular Formula C8H9ClN2
Molecular Weight (g/mol) 168.624
MDL Number MFCD01861472
InChI Key QREZLLYPLRPULF-UHFFFAOYSA-N
Synonym 4-aminomethyl benzonitrile hydrochloride, 4-cyanobenzylamine hcl, 4-cyanobenzylamine hydrochloride, 4-aminomethyl benzonitrile hcl, 4-aminomethyl-benzonitrile hydrochloride, benzonitrile, 4-aminomethyl-, monohydrochloride, p-aminomethyl benzonitrile hydrochloride, pubchem2057, acmc-209dkg, 4-aminomethylbenzonitrile hcl
PubChem CID 9942250
IUPAC Name 4-(aminomethyl)benzonitrile;hydrochloride
SMILES C1=CC(=CC=C1CN)C#N.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 168.62
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Aminomethyl)benzonitrile Hydrochloride