missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)benzeneboronic acid pinacol ester hydrochloride, 95%

Catalog No. AAH52691MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52691MD 250 mg
AAH5269103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52691MD Supplier Thermo Scientific Chemicals Supplier No. H52691MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 850568-55-7 | C13H21BClNO2 | 269.576 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 850568-55-7
Molecular Formula C13H21BClNO2
Molecular Weight (g/mol) 269.576
MDL Number MFCD02179455
InChI Key KPECMJIHZZWTJN-UHFFFAOYSA-N
Synonym 4-aminomethylphenylboronic acid, pinacol ester, hcl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hydrochloride, 4-aminomethylphenylboronic acid pinacol ester hydrochloride, 4-aminomethylphenylboronic acid pinacol ester hcl, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hydrochloride, benzenemethanamine,4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylamine hydrochloride, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hcl, 4-aminomethylohenylboronic acid pinacl ester hcl
PubChem CID 16427088
IUPAC Name [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanamine;hydrochloride
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN.Cl

Specifications

Melting Point 228°C to 238°C
Quantity 250 mg
Formula Weight 269.58
Percent Purity 95%
Chemical Name or Material 4-(Aminomethyl)benzeneboronic acid pinacol ester hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only