missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzylamine 98.0+%, TCI America™

Catalog No. A143625G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A143625G 25 g
A1436250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A143625G Supplier TCI America Supplier No. A143625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4403-71-8
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00075513
InChI Key BFWYZZPDZZGSLJ-UHFFFAOYSA-N
Synonym 4-aminobenzylamine, 4-aminomethyl aniline, p-aminobenzylamine, benzenemethanamine, 4-amino, 4-aminomethyl-phenylamine, 4-aminomethyl phenylamine, 4-aminobenzyl amine dihydrochloride, 4-amonobenzylae, p-aminobenzylamin, 4-amino benzylamine
PubChem CID 427814
IUPAC Name 4-(aminomethyl)aniline
SMILES C1=CC(=CC=C1CN)N

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3259
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Aminobenzylamine
EINECSNumber (3)-3897
TSCA No