missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzotrifluoride 98.0+%, TCI America™

Catalog No. A066225ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A066225ML 25 mL
A0662100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A066225ML Supplier TCI America Supplier No. A066225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 455-14-1
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.127
MDL Number MFCD00064396
InChI Key ODGIMMLDVSWADK-UHFFFAOYSA-N
Synonym 4-trifluoromethyl aniline, 4-aminobenzotrifluoride, p-aminobenzotrifluoride, p-trifluoromethylaniline, benzenamine, 4-trifluoromethyl, 4-trifluoromethyl benzenamine, p-trifluoromethyl aniline, aniline, p-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluidine, a,a,a-trifluoro-p-toluidine
PubChem CID 9964
ChEBI CHEBI:40750
IUPAC Name 4-(trifluoromethyl)aniline
SMILES C1=CC(=CC=C1C(F)(F)F)N

Specifications

Boiling Point 117°C
Quantity 25 mL
UN Number 2810
Formula Weight 161.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Aminobenzotrifluoride
EINECSNumber (3)-0223
RTECSNumber XU9260000
TSCA Yes
Recommended Storage Refrigerator